siRNA Details
Gene Symbol: 2010002M12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053217.2 | NP_444447.1 | 3 | 602 | ||
GenBank mRNA | AK008052.1 | - | 2 | 454 | ||
AK136525.1 | - | 2 | 454 | |||
AK153247.1 | - | 3 | 603 | |||
BC138202.1 | - | 2 | 433 | 8861221 |
Gene Symbol: Gm14446 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101605.1 | NP_001095075.1 | 2 | 448 | ||
NM_001110517.1 | NP_001103987.1 | 2 | 529 | |||
GenBank mRNA | AK090072.1 | - | 2 | 525 |
Gene Symbol: Ifit1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008331.3 | NP_032357.2 | 2 | 471 | ||
GenBank mRNA | AK089608.1 | - | 2 | 478 | ||
AK131669.1 | - | 2 | 468 | |||
AK144230.1 | - | 2 | 476 | |||
AK150352.1 | - | 2 | 478 | |||
AK150424.1 | - | 2 | 472 | |||
AK150609.1 | - | 2 | 481 | |||
AK151125.1 | - | 2 | 481 | |||
AK151158.1 | - | 2 | 472 | |||
AK151278.1 | - | 2 | 481 | |||
AK151298.1 | - | 2 | 475 | |||
AK151983.1 | - | 2 | 474 | |||
AK152112.1 | - | 2 | 474 | |||
AK152853.1 | - | 1 | 146 | |||
AK153184.1 | - | 2 | 474 | |||
AK153500.1 | - | 2 | 472 | |||
AK153532.1 | - | 2 | 481 | |||
AK157848.1 | - | 2 | 472 | |||
BC003768.1 | - | 2 | 455 | 3496673 | ||
CT010226.1 | - | 1 | 377 | |||
U43084.1 | - | 2 | 446 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
