siRNA Details
Gene Symbol: Il18r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161842.1 | NP_001155314.1 | 4 | 535 | ||
NM_001161843.1 | NP_001155315.1 | 4 | 407 | |||
NM_008365.2 | NP_032391.1 | 3 | 460 | |||
GenBank mRNA | AK087129.1 | - | 3 | 409 | ||
AK089237.1 | - | 4 | 536 | |||
AK170705.1 | - | 4 | 496 | |||
BC020296.1 | - | 3 | 222 | |||
BC023240.1 | - | 4 | 376 | 4483261 | ||
U43673.1 | - | 3 | 460 |
