siRNA Details
Gene Symbol: TRPM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301212.1 | NP_001288141.1 | 3 | 384 | ||
NM_017672.5 | NP_060142.3 | 3 | 384 | |||
XM_005254486.3 | XP_005254543.1 | 3 | 379 | |||
XM_017022350.1 | XP_016877839.1 | 3 | 144 | |||
XM_017022351.1 | XP_016877840.1 | 3 | 144 | |||
XM_017022352.1 | XP_016877841.1 | 3 | 144 | |||
XM_017022353.1 | XP_016877842.1 | 3 | 144 | |||
XM_017022354.1 | XP_016877843.1 | 3 | 209 | |||
XM_017022356.1 | XP_016877845.1 | 3 | 144 | |||
GenBank mRNA | AF346629.2 | - | 3 | 340 | ||
AK000124.1 | - | 3 | 315 | |||
AY032950.1 | - | 3 | 373 | |||
BC128987.1 | - | 3 | 195 | |||
BC128988.1 | - | 3 | 195 | |||
BP354541.1 | - | 3 | 384 | |||
DB127039.1 | - | 3 | 366 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
