siRNA Details
Gene Symbol: HP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126102.2 | NP_001119574.1 | 5 | 1190 | ||
NM_001318138.1 | NP_001305067.1 | 5 | 1190 | |||
NM_005143.4 | NP_005134.1 | 7 | 1367 | |||
GenBank mRNA | AK055872.1 | - | 3 | 2945 | ||
BC017862.1 | - | 6 | 1121 | 4700637 | ||
BC058031.1 | - | 5 | 1099 | 4723094 | ||
BC070299.1 | - | 4 | 1029 | 4716454 | ||
BC107587.1 | - | 5 | 1163 | 4734599 | ||
BC121124.1 | - | 5 | 1153 | 40120682 | ||
BC121125.1 | - | 7 | 1330 | 40120683 | ||
K00422.1 | - | 7 | 1318 | |||
K01763.1 | - | 5 | 1155 | |||
L29394.1 | - | 7 | 1332 | |||
T71743.1 | - | 1 | 250 | |||
X00637.1 | - | 5 | 1143 |
Gene Symbol: HPR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020995.3 | NP_066275.3 | 5 | 1162 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
