siRNA Details
Gene Symbol: H2-DMb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010387.3 | NP_034517.2 | 6 | 1244 | ||
GenBank mRNA | AK032745.1 | - | 6 | 1245 | ||
BC002237.1 | - | 1205 | 3491702 | |||
BC003718.1 | - | 6 | 1184 | 3486844 | ||
X62743.1 | - | 1144 |
