siRNA Details
Gene Symbol: Hoxb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010459.7 | NP_034589.3 | 2 | 1649 | ||
GenBank mRNA | AK133945.1 | - | 2 | 1178 | ||
M36654.1 | - | 2 | 1215 |

More Information
Other Species Targeted

Gene Ontology Categories:
