siRNA Details
Gene Symbol: Epb4.1l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199265.1 | NP_001186194.1 | 2 | 455 | ||
NM_013511.3 | NP_038539.2 | 2 | 346 | |||
GenBank mRNA | AB370258.1 | - | 1 | 257 | ||
AB370259.1 | - | 1 | 257 | |||
AB370260.1 | - | 1 | 257 | |||
AB370263.1 | - | 1 | 257 | |||
AF044312.1 | - | 1 | 257 | |||
AJ539142.1 | - | 1 | 257 | |||
AJ539143.1 | - | 1 | 257 | |||
AJ539144.1 | - | 1 | 257 | |||
AJ542537.1 | - | 1 | 257 | |||
AK043148.1 | - | 1 | 234 | |||
AK052475.1 | - | 2 | 346 | |||
AK144088.1 | - | 2 | 456 | |||
AK167434.1 | - | 2 | 372 | |||
AK170275.1 | - | 2 | 398 | |||
BC023220.1 | - | 2 | 342 | |||
BC023768.1 | - | 2 | 368 | |||
BC026708.1 | - | 2 | 327 | |||
BC029938.1 | - | 2 | 484 | |||
KF534728.1 | - | 1 | 257 |
