siRNA Details
Gene Symbol: Ap1g2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF068707.1 | - | 21 | 2503 | ||
AK052384.1 | - | 20 | 2656 | |||
AK147177.1 | - | 22 | 2541 | |||
BC058402.1 | - | 20 | 2547 | |||
BC080798.1 | - | 21 | 2449 | |||
BC108374.1 | - | 22 | 2528 | 4936738 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
