siRNA Details
Gene Symbol: Atm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007499.2 | NP_031525.2 | 5 | 554 | ||
GenBank mRNA | AK139863.1 | - | 5 | 558 | ||
BC138525.1 | - | 4 | 456 | 8861545 | ||
BY725995.1 | - | 5 | 554 | |||
U43678.1 | - | 4 | 456 |
