siRNA Details
Gene Symbol: Atm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007499.2 | NP_031525.2 | 6 | 792 | ||
GenBank mRNA | AK139863.1 | - | 6 | 796 | ||
BC138525.1 | - | 5 | 694 | 8861545 | ||
U43678.1 | - | 5 | 694 |

More Information
Other Species Targeted

Gene Ontology Categories:
