siRNA Details
Gene Symbol: Bmp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC100751.1 | - | 1 | 286 | ||
BC100752.1 | - | 1 | 286 | |||
BC100754.1 | - | 1 | 286 | |||
BC141283.1 | - | 1 | 818 | 9055895 | ||
L41145.1 | - | 1 | 287 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
