siRNA Details
Gene Symbol: Calca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007587.2 | NP_031613.2 | 4 | 447 | ||
GenBank mRNA | AF078167.1 | - | 438 | |||
AK132582.1 | - | 4 | 397 | |||
BC030071.1 | - | 4 | 378 | 1399214 | ||
X97991.1 | - | 4 | 408 |

More Information
Other Species Targeted

Gene Ontology Categories:
