siRNA Details
Gene Symbol: Calca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007587.2 | NP_031613.2 | 4 | 729 | ||
GenBank mRNA | AK132582.1 | - | 4 | 679 | ||
BC030071.1 | - | 4 | 660 | 1399214 | ||
X97991.1 | - | 4 | 690 |

More Information
Other Species Targeted

Gene Ontology Categories:
