siRNA Details
Gene Symbol: Capn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301250.1 | NP_001288179.1 | 5 | 737 | ||
NM_007602.4 | NP_031628.1 | 5 | 901 | |||
GenBank mRNA | AK155179.1 | - | 5 | 879 | ||
AK164293.1 | - | 5 | 811 | |||
AK170034.1 | - | 5 | 838 | |||
BC014767.1 | - | 5 | 729 | 4458488 | ||
U85020.1 | - | 5 | 820 | |||
Y10656.1 | - | 5 | 833 |
