siRNA Details
Gene Symbol: Capn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301250.1 | NP_001288179.1 | 13 | 2619 | ||
NM_007602.4 | NP_031628.1 | 13 | 2783 | |||
GenBank mRNA | AK139476.1 | - | 2 | 932 | ||
AK155179.1 | - | 13 | 2761 | |||
AK170034.1 | - | 13 | 2720 | |||
U85020.1 | - | 13 | 2702 |
