siRNA Details
Gene Symbol: EFNA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005227.2 | NP_005218.1 | 3, 4 | 557 | ||
GenBank mRNA | AJ006352.1 | - | 3, 4 | 497 | ||
BC107483.1 | - | 3, 4 | 547 | 40030599 | ||
BE780161.1 | - | 2, 3 | 111 | |||
CD671399.1 | - | 3, 4 | 563 | |||
CR533569.1 | - | 3, 4 | 470 | |||
U14188.1 | - | 3, 4 | 497 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
