siRNA Details
Gene Symbol: Cd36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159555.1 | NP_001153027.1 | 15 | 1932 | ||
NM_001159556.1 | NP_001153028.1 | 15 | 1792 | |||
NM_001159557.1 | NP_001153029.1 | 16 | 1843 | |||
NM_001159558.1 | NP_001153030.1 | 16 | 1959 | |||
NM_007643.4 | NP_031669.3 | 15 | 1935 | |||
GenBank mRNA | AK004192.1 | - | 3 | 254 | ||
AK052825.1 | - | 15 | 1736 | |||
AK128908.1 | - | 16 | 1736 | |||
AK150648.1 | - | 15 | 1756 | |||
AK150865.1 | - | 11 | 1128 | |||
AK151355.1 | - | 15 | 1771 | |||
AK152642.1 | - | 10 | 927 | |||
AK152904.1 | - | 13 | 1442 | |||
AK153095.1 | - | 10 | 1045 | |||
AK172162.1 | - | 16 | 1781 | |||
BC010262.1 | - | 15 | 1744 | 3481681 |
