siRNA Details
Gene Symbol: Scarb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007644.3 | NP_031670.1 | 3 | 646 | ||
GenBank mRNA | AB008553.1 | - | 3 | 551 | ||
AK011123.1 | - | 3 | 576 | |||
AK083038.1 | - | 3 | 647 | |||
AK144235.1 | - | 3 | 678 | |||
AK149102.1 | - | 592 | ||||
BC029073.1 | - | 3 | 554 | 3962669 |
