siRNA Details
Gene Symbol: Scarb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007644.3 | NP_031670.1 | 3 | 653 | ||
GenBank mRNA | AB008553.1 | - | 3 | 558 | ||
AK011123.1 | - | 3 | 583 | |||
AK083038.1 | - | 3 | 654 | |||
AK144235.1 | - | 3 | 685 | |||
AK149102.1 | - | 599 | ||||
BC029073.1 | - | 3 | 561 | 3962669 |
