siRNA Details
Gene Symbol: GLUD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005271.4 | NP_005262.1 | 1, 2 | 727 | ||
GenBank mRNA | AK094782.1 | - | 3, 4 | 391 | ||
AK222818.1 | - | 1, 2 | 521 | |||
AK297441.1 | - | 2, 3 | 372 | |||
AK297456.1 | - | 2, 3 | 408 | |||
AK303458.1 | - | 2, 3 | 373 | |||
AK311722.1 | - | 2, 3 | 454 | |||
BC040132.1 | - | 1, 2 | 500 | 5740251 | ||
BC112946.1 | - | 1, 2 | 510 | 6138490 | ||
GU727646.1 | - | 1, 2 | 449 | |||
J03248.1 | - | 1, 2 | 512 | |||
M20867.1 | - | 1, 2 | 546 | |||
M37154.1 | - | 1, 2 | 462 | |||
X07674.1 | - | 1, 2 | 462 | |||
X07769.1 | - | 1, 2 | 462 |
Gene Symbol: GLUD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012084.3 | NP_036216.2 | 1 | 526 | ||
GenBank mRNA | AK313356.1 | - | 1 | 523 | ||
BC050732.2 | - | 1 | 526 | 6164919 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
