siRNA Details
Gene Symbol: Ccr10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007721.4 | NP_031747.2 | 2 | 1404 | ||
GenBank mRNA | AF215983.1 | - | 2 | 1380 | ||
AK090044.1 | - | 2 | 1407 | |||
BC117043.1 | - | 2 | 1407 | 40126362 | ||
BC120542.1 | - | 2 | 1407 | 40129465 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
