siRNA Details
Gene Symbol: Dbi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037999.2 | NP_001033088.1 | 2 | 260 | ||
NM_007830.4 | NP_031856.1 | 2 | 254 | |||
GenBank mRNA | AK008540.1 | - | 2 | 175 | ||
AK008576.1 | - | 2 | 175 | |||
AK018720.1 | - | 2 | 176 | |||
AK020692.1 | - | 2 | 224 | |||
AK027906.1 | - | 2 | 176 | |||
AK078428.1 | - | 2 | 237 | |||
AK145278.1 | - | 2 | 176 | |||
AK148753.1 | - | 2 | 176 | |||
AM000002.1 | - | 2 | 260 | |||
AV001592.1 | - | 2 | 148 |

More Information
Other Species Targeted

Gene Ontology Categories:
