siRNA Details
Gene Symbol: Dcc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007831.3 | NP_031857.2 | 2 | 958 | ||
GenBank mRNA | AK049746.1 | - | 2 | 932 | ||
AK090377.1 | - | 2 | 959 | |||
AK157715.1 | - | 2 | 907 | |||
X85788.1 | - | 508 |
