siRNA Details
Gene Symbol: Gm14851 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177482.1 | NP_001170953.1 | 2 | 344 | ||
GenBank mRNA | AV063918.1 | - | 1 | 179 | ||
AV064248.1 | - | 1 | 180 |
Gene Symbol: AY761184 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007582.3 | NP_001007583.1 | 2 | 389 | ||
GenBank mRNA | AK008498.1 | - | 2 | 395 | ||
EU760895.1 | - | 1 | 185 |
Gene Symbol: Defa-rs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007844.2 | NP_031870.2 | 2 | 389 | ||
GenBank mRNA | AK008584.1 | - | 2 | 390 | ||
BC117042.1 | - | 2 | 364 | 40126361 | ||
BC117044.1 | - | 2 | 364 | 40126363 | ||
BC145556.1 | - | 2 | 394 | 9054120 | ||
BC171966.1 | - | 2 | 394 | 9054252 | ||
EU760893.1 | - | 1 | 185 | |||
EU760894.1 | - | 1 | 185 | |||
M33226.1 | - | 2 | 369 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
