siRNA Details
Gene Symbol: Usp17la | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007887.2 | NP_031913.1 | 3 | 1955 | ||
GenBank mRNA | U41636.1 | - | 3 | 1967 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
