siRNA Details
Gene Symbol: Anp32a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009672.3 | NP_033802.2 | 3 | 439 | ||
GenBank mRNA | AF022957.1 | - | 3 | 348 | ||
AK020753.1 | - | 3 | 401 | |||
BC062899.1 | - | 3 | 354 | 6415168 | ||
U73478.1 | - | 3 | 306 |
Gene Symbol: Anp32-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY702092.1 | - | 1 | 281 |

More Information
Other Species Targeted

Gene Ontology Categories:
