siRNA Details
Gene Symbol: Apex1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009687.2 | NP_033817.1 | 5 | 1212 | ||
GenBank mRNA | AK013296.1 | - | 5 | 1180 | ||
BC052401.1 | - | 5 | 1183 | 5715485 | ||
D90374.1 | - | 4 | 1140 |

More Information
Other Species Targeted

Gene Ontology Categories:
