siRNA Details
Gene Symbol: Nr2f2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009697.3 | NP_033827.2 | 3 | 2194 | ||
NM_183261.3 | NP_899084.2 | 3 | 2142 | |||
GenBank mRNA | AK032882.1 | - | 1 | 1266 | ||
AK034619.1 | - | 2 | 1314 | |||
AK140125.1 | - | 3 | 2118 | |||
BC042484.1 | - | 3 | 2194 | 5368085 | ||
X76653.1 | - | 3 | 1963 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
