siRNA Details
Gene Symbol: GLUD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005271.4 | NP_005262.1 | 1, 2 | 731 | ||
GenBank mRNA | AK094782.1 | - | 3, 4 | 395 | ||
AK222818.1 | - | 1, 2 | 525 | |||
AK297441.1 | - | 2, 3 | 376 | |||
AK297456.1 | - | 2, 3 | 412 | |||
AK303458.1 | - | 2, 3 | 377 | |||
AK311722.1 | - | 2, 3 | 458 | |||
BC040132.1 | - | 1, 2 | 504 | 5740251 | ||
BC112946.1 | - | 1, 2 | 514 | 6138490 | ||
GU727646.1 | - | 1, 2 | 453 | |||
J03248.1 | - | 1, 2 | 516 | |||
M20867.1 | - | 1, 2 | 550 | |||
M37154.1 | - | 1, 2 | 466 | |||
X07674.1 | - | 1, 2 | 466 | |||
X07769.1 | - | 1, 2 | 466 |
Gene Symbol: GLUD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012084.3 | NP_036216.2 | 1 | 530 | ||
GenBank mRNA | AK313356.1 | - | 1 | 527 | ||
BC050732.2 | - | 1 | 530 | 6164919 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
