siRNA Details
Gene Symbol: GPT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005309.2 | NP_005300.1 | 9 | 1416 | ||
XM_011516993.2 | XP_011515295.1 | 10 | 2106 | |||
GenBank mRNA | AK225562.1 | - | 9 | 1416 | ||
BC018207.1 | - | 10 | 1350 | 4179699 | ||
BT006992.1 | - | 9 | 1193 | |||
D10355.1 | - | 9 | 1184 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
