siRNA Details
Gene Symbol: MAP2K5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206804.1 | NP_001193733.1 | 13 | 885 | ||
NM_002757.3 | NP_002748.1 | 13 | 1469 | |||
NM_145160.2 | NP_660143.1 | 13 | 1469 | |||
XM_011521787.2 | XP_011520089.1 | 13 | 1546 | |||
XM_011521788.2 | XP_011520090.1 | 13 | 1546 | |||
GenBank mRNA | AK024766.1 | - | 2 | 63 | ||
AK057521.1 | - | 7 | 597 | |||
AK293459.1 | - | 13 | 885 | |||
AK315435.1 | - | 13 | 1016 | |||
BC008838.2 | - | 13 | 1126 | 4100679 | ||
BT006780.1 | - | 13 | 816 | |||
CR542229.1 | - | 13 | 816 | |||
U71087.1 | - | 13 | 877 | |||
U71088.2 | - | 13 | 821 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
