siRNA Details
Gene Symbol: Cd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037801.2 | NP_001032890.1 | 4 | 685 | ||
NM_009852.3 | NP_033982.3 | 4 | 685 | |||
GenBank mRNA | AK030822.1 | - | 4 | 655 | ||
AK030823.1 | - | 4 | 686 | |||
AK088050.1 | - | 4 | 663 | |||
AK141213.1 | - | 1 | 135 | |||
U12434.1 | - | 4 | 483 | |||
U35370.1 | - | 4 | 601 | |||
U37543.1 | - | 4 | 642 | |||
U37544.1 | - | 4 | 642 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
