siRNA Details
Gene Symbol: Cd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037801.2 | NP_001032890.1 | 12 | 2549 | ||
NM_009852.3 | NP_033982.3 | 13 | 2666 | |||
GenBank mRNA | AK030822.1 | - | 13 | 2636 | ||
AK030823.1 | - | 13 | 2667 | |||
AK088050.1 | - | 12 | 2533 | |||
AK141213.1 | - | 9 | 1813 | |||
U12434.1 | - | 12 | 2347 | |||
U35370.1 | - | 12 | 2465 | |||
U37543.1 | - | 13 | 2627 | |||
U37544.1 | - | 12 | 2164 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
