siRNA Details
Gene Symbol: Cfhr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025575.2 | NP_001020746.1 | 3 | 333 | ||
GenBank mRNA | AJ565908.1 | - | 3 | 282 | ||
AK149511.1 | - | 3 | 324 | |||
M29007.1 | - | 325 |
Gene Symbol: Cfh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009888.3 | NP_034018.2 | 8 | 1167 | ||
GenBank mRNA | AK087707.1 | - | 8 | 1086 | ||
BC066092.1 | - | 8 | 1036 | 6842070 | ||
M12660.1 | - | 8 | 1078 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
