siRNA Details
Gene Symbol: H3F3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005324.4 | NP_005315.1 | 4 | 895 | ||
GenBank mRNA | AF218029.1 | - | 4 | 719 | ||
AK130772.1 | - | 4 | 738 | |||
AK304607.1 | - | 4 | 670 | |||
BC001124.2 | - | 4 | 723 | 2988279 | ||
BC006497.2 | - | 4 | 723 | 2959756 | ||
BC012813.2 | - | 4 | 723 | 2959756 | ||
BC017558.1 | - | 4 | 727 | 3910031 | ||
BC020466.1 | - | 2 | 431 | |||
BC108701.1 | - | 4 | 702 | 6144255 | ||
BX537379.1 | - | 4 | 740 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
