siRNA Details
Gene Symbol: Col6a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243008.1 | NP_001229937.1 | 7 | 2948 | ||
NM_001243009.1 | NP_001229938.1 | 4 | 1127 | |||
GenBank mRNA | AF034136.1 | - | 7 | 2905 | ||
AF064749.1 | - | 4 | 934 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
