siRNA Details
Gene Symbol: Copa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009938.4 | NP_034068.3 | 18 | 2111 | ||
GenBank mRNA | AK043479.1 | - | 18 | 1837 | ||
BC024070.1 | - | 18 | 1764 | 5097228 | ||
BC047429.1 | - | 18 | 1802 | 3498656 | ||
BC082785.1 | - | 14 | 1451 |
