siRNA Details
Gene Symbol: HSP90AA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017963.2 | NP_001017963.2 | 6 | 1640 | ||
NM_005348.3 | NP_005339.3 | 5 | 1119 | |||
XM_011536718.2 | XP_011535020.1 | 6 | 1637 | |||
GenBank mRNA | AF028832.1 | - | 1 | 804 | ||
AJ890082.1 | - | 6 | 1577 | |||
AJ890083.1 | - | 5 | 969 | |||
AK056446.1 | - | 6 | 1577 | |||
AK129557.1 | - | 5 | 767 | |||
AK291115.1 | - | 5 | 988 | |||
AK291607.1 | - | 5 | 988 | |||
AK310595.1 | - | 605 | ||||
BC000987.2 | - | 2 | 379 | |||
BC023006.1 | - | 3 | 647 | |||
BC108695.1 | - | 4 | 975 | |||
BC121062.2 | - | 5 | 989 | 40118488 | ||
BX247955.1 | - | 3 | 515 | |||
GQ149083.1 | - | 5 | 1119 | |||
GQ472230.1 | - | 5 | 989 | |||
X15183.1 | - | 5 | 989 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
