siRNA Details
Gene Symbol: Pcdha7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009957.1 | NP_034087.1 | 1 | 951 | ||
GenBank mRNA | AB008179.1 | - | 1 | 844 | ||
AY013765.1 | - | 1 | 951 | |||
BC080863.1 | - | 1 | 1073 | 30356469 |
Gene Symbol: Pcdha7-g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301259.1 | NP_001288188.1 | 1 | 1073 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
