siRNA Details
Gene Symbol: MB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK300657.1 | - | 3 | 217 | ||
AK314057.1 | - | 3 | 320 | |||
BC014547.1 | - | 2 | 255 | 4244838 | ||
CB140824.1 | - | 3 | 364 | |||
CR456516.1 | - | 2 | 227 | |||
CR541949.1 | - | 2 | 179 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
