siRNA Details
Gene Symbol: METTL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005371.5 | NP_005362.3 | 6 | 923 | ||
NM_023033.3 | NP_075422.3 | 5 | 738 | |||
XM_005268873.2 | XP_005268930.1 | 7 | 1023 | |||
GenBank mRNA | AK223202.1 | - | 5 | 712 | ||
X96698.1 | - | 6 | 894 | |||
Y18643.1 | - | 6 | 923 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
