siRNA Details
Gene Symbol: Cxadr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009988.4 | NP_034118.1 | 8 | 1236 | ||
GenBank mRNA | AK160457.1 | - | 8 | 1158 | ||
BC016457.1 | - | 8 | 1151 | 4216727 | ||
U90715.1 | - | 8 | 1136 | |||
Y11929.1 | - | 8 | 1190 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
