siRNA Details
Gene Symbol: Cyp2a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007812.4 | NP_031838.2 | 9 | 1632 | ||
GenBank mRNA | AK132617.1 | - | 9 | 1631 | ||
AK149439.1 | - | 9 | 1633 | |||
BC046605.1 | - | 9 | 1606 | |||
BC058743.1 | - | 9 | 1604 | |||
BC138796.1 | - | 9 | 1628 | 8861818 | ||
BC138798.1 | - | 9 | 1628 | 8861820 |
Gene Symbol: Cyp2a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009997.2 | NP_034127.2 | 9 | 1633 | ||
GenBank mRNA | BC010761.1 | - | 9 | 1624 | 4239813 | |
BC011233.1 | - | 1609 | 4190317 | |||
BC063778.1 | - | 9 | 1656 | 5044482 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
