siRNA Details
Gene Symbol: Dab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010014.3 | NP_034144.1 | 7 | 936 | ||
GenBank mRNA | AK005640.1 | - | 6 | 747 | ||
Y08381.1 | - | 7 | 884 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
