siRNA Details
Gene Symbol: Dsg1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK036986.1 | - | 15 | 3866 | ||
AK076540.1 | - | 1 | 194 |
Gene Symbol: Dsg1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010079.2 | NP_034209.2 | 15 | 3560 | ||
GenBank mRNA | AK029294.1 | - | 1 | 1219 | ||
AK037234.1 | - | 1 | 433 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
