siRNA Details
Gene Symbol: Adam26a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010085.2 | NP_034215.2 | 2 | 1482 | ||
GenBank mRNA | AF167404.1 | - | 2 | 1441 | ||
AK029756.1 | - | 2 | 1483 |
Gene Symbol: Adam26b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009547.2 | NP_001009547.1 | 2 | 1472 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
