siRNA Details
Gene Symbol: Prl8a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289919.1 | NP_001276848.1 | 6 | 658 | ||
NM_010088.2 | NP_034218.1 | 6 | 661 | |||
GenBank mRNA | AF011385.1 | - | 6 | 645 | ||
AF015729.1 | - | 6 | 655 | |||
AK131668.1 | - | 6 | 659 | |||
AK146049.1 | - | 6 | 662 | |||
BC061150.1 | - | 6 | 658 | 30317747 |
