siRNA Details
Gene Symbol: Casp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277932.1 | NP_001264861.1 | 8 | 2328 | ||
NM_015733.5 | NP_056548.2 | 9 | 2438 | |||
GenBank mRNA | AK085152.1 | - | 9 | 2195 | ||
AK154495.1 | - | 9 | 2209 | |||
BC056372.1 | - | 9 | 2220 | 6834067 | ||
BC056447.1 | - | 9 | 2438 | 5721188 |
