siRNA Details
Gene Symbol: Vcan | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081249.1 | NP_001074718.1 | 4 | 930 | ||
NM_001134474.1 | NP_001127946.1 | 4 | 930 | |||
NM_001134475.1 | NP_001127947.1 | 4 | 930 | |||
NM_019389.2 | NP_062262.2 | 4 | 930 | |||
NM_172955.1 | NP_766543.1 | 4 | 930 | |||
GenBank mRNA | AK014525.1 | - | 4 | 740 | ||
AK034871.1 | - | 4 | 737 | |||
AK040503.1 | - | 1 | 64 | |||
AK135324.1 | - | 4 | 927 | |||
AK163927.1 | - | 4 | 787 | |||
AK165069.1 | - | 4 | 733 | |||
BC021652.1 | - | 4 | 770 | |||
BC096495.1 | - | 4 | 740 | |||
D16263.1 | - | 3 | 490 | |||
D28599.1 | - | 4 | 662 | |||
D32040.1 | - | 4 | 662 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
