siRNA Details
Gene Symbol: KHK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000221.2 | NP_000212.1 | 8 | 2212 | ||
NM_006488.2 | NP_006479.1 | 8 | 2212 | |||
XM_005264294.3 | XP_005264351.1 | 9 | 2355 | |||
XM_005264296.3 | XP_005264353.1 | 8 | 2198 | |||
XM_005264298.3 | XP_005264355.1 | 6 | 1965 | |||
XM_017004060.1 | XP_016859549.1 | 9 | 2352 | |||
XM_017004061.1 | XP_016859550.1 | 7 | 2082 | |||
GenBank mRNA | AK130033.1 | - | 8 | 1729 | ||
BX648873.1 | - | 6 | 1479 | |||
X78678.1 | - | 8 | 1703 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
